missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asialofetuin Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
£160.00 - £360.00
Specifications
| Antigen | Asialofetuin |
|---|---|
| Concentration | 5.0 mg/mL |
| Dilution | Simple Western, ELISA 1:1000-1:4000 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18051506
|
Novus Biologicals
NB100-62357-0.1ML |
0.1 mL |
£160.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18768253
|
Novus Biologicals
NB100-62357 |
0.5 mL |
£360.00
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Asialofetuin Polyclonal specifically detects Asialofetuin in Bovine samples. It is validated for Simple Western, ELISA.Specifications
| Asialofetuin | |
| Simple Western, ELISA 1:1000-1:4000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline with 0.09% Sodium Azide | |
| AHSG | |
| IgG | |
| Protein G purified | |
| Asialofetuin 100% Fetuin (ab at 1/2000) 100% Fetuin (ab at 1/8000) 50% |
| 5.0 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Sheep | |
| Bovine | |
| P12763 | |
| Native | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title