missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asialofetuin Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB100-62357
This item is not returnable.
View return policy
Description
Asialofetuin Polyclonal specifically detects Asialofetuin in Bovine samples. It is validated for Simple Western, ELISA.
Specifications
| Asialofetuin | |
| Polyclonal | |
| Unconjugated | |
| Phosphate buffered saline with 0.09% Sodium Azide | |
| AHSG | |
| Sheep | |
| Protein G purified | |
| RUO | |
| Asialofetuin 100% Fetuin (ab at 1/2000) 100% Fetuin (ab at 1/8000) 50% | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| 5.0 mg/mL | |
| Simple Western, ELISA 1:1000-1:4000 | |
| P12763 | |
| AHSG | |
| Native | |
| 0.5 mL | |
| Primary | |
| Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction