missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ASCC2 Monoclonal Antibody (24GB6350)
GREENER_CHOICE

Mouse Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen ASCC2
Clone 24GB6350
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275385
GREENER_CHOICE
View Documents
Invitrogen™
MA557345
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30274923
missing translation for 'viewDocuments'
Invitrogen™
MA557346
20 μL
£172.00
20µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ASCC2 Monoclonal Antibody for Western Blot

Plays a role in DNA damage repair as component of the ASCC complex. Recruits ASCC3 and ALKBH3 to sites of DNA damage by binding to polyubiquitinated proteins that have 'Lys-63'-linked polyubiquitin chains (PubMed:29144457). Part of the ASC-1 complex that enhances NF-kappa-B, SRF and AP1 transactivation (PubMed:12077347). [UniProt]
Specifications

Specifications

ASCC2
-20°C
Monoclonal
Liquid
IgG1
Human, Mouse, Rat
ASCC2
1700011I11Rik; 2610034L15Rik; activating signal cointegrator 1 complex subunit 2; AI482016; ASC-1 complex subunit P100; Asc1p100; ASCC2; AW046480; p100; RGD1561422; trip4 complex subunit p100
ASCC2
Primary
Affinity chromatography
24GB6350
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Mouse
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q91WR3, Q9H1I8
498402, 75452, 84164
Recombinant protein of human ASCC2 (aa 1-757).
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.