missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASCC2 Monoclonal Antibody (24GB6350)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA557345
This item is not returnable.
View return policy
Description
ASCC2 Monoclonal Antibody for Western Blot
Plays a role in DNA damage repair as component of the ASCC complex. Recruits ASCC3 and ALKBH3 to sites of DNA damage by binding to polyubiquitinated proteins that have 'Lys-63'-linked polyubiquitin chains (PubMed:29144457). Part of the ASC-1 complex that enhances NF-kappa-B, SRF and AP1 transactivation (PubMed:12077347). [UniProt].
Specifications
| ASCC2 | |
| Monoclonal | |
| Unconjugated | |
| ASCC2 | |
| 1700011I11Rik; 2610034L15Rik; activating signal cointegrator 1 complex subunit 2; AI482016; ASC-1 complex subunit P100; Asc1p100; ASCC2; AW046480; p100; RGD1561422; trip4 complex subunit p100 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 498402, 75452, 84164 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB6350 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q91WR3, Q9H1I8 | |
| ASCC2 | |
| Recombinant protein of human ASCC2 (aa 1-757). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction