missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP42 Antibody (2F1A7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | ARHGAP42 |
|---|---|
| Clone | 2F1A7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18253431
|
Novus Biologicals
NBP2-61675 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677888
|
Novus Biologicals
NBP2-61675-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARHGAP42 Monoclonal specifically detects ARHGAP42 in Human samples. It is validated for Western Blot, ELISA.Specifications
| ARHGAP42 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| 143872 | |
| Purified recombinant fragment of human ARHGAP42 (AA: 577-719) expressed in E. Coli. | |
| Primary | |
| 98.6 kDa |
| 2F1A7 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| FLJ32810, Rho GTPase activating protein 42 | |
| ARHGAP42 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title