missing translation for 'onlineSavingsMsg'
Learn More

ARHGAP42 Antibody (2F1A7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-61675

 View more versions of this product

Product Code. 18253431

  • £435.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ARHGAP42 Monoclonal specifically detects ARHGAP42 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ARHGAP42
Monoclonal
1 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000
FLJ32810, Rho GTPase activating protein 42
Mouse
98.6 kDa
0.1 mL
143872
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, ELISA
2F1A7
Unconjugated
PBS with 0.05% Sodium Azide
ARHGAP42
Purified recombinant fragment of human ARHGAP42 (AA: 577-719) expressed in E. Coli.
Protein G purified
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.