missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP42 Antibody (2F1A7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61675-0.025ml
This item is not returnable.
View return policy
Description
ARHGAP42 Monoclonal specifically detects ARHGAP42 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| ARHGAP42 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| FLJ32810, Rho GTPase activating protein 42 | |
| Mouse | |
| 98.6 kDa | |
| 0.025 mL | |
| 143872 | |
| Purified |
| Western Blot, ELISA | |
| 2F1A7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ARHGAP42 | |
| Purified recombinant fragment of human ARHGAP42 (AA: 577-719) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction