missing translation for 'onlineSavingsMsg'
Learn More

TCR β chain Hamster anti-Mouse, Biotin, Clone: H57-597, BD

Hamster Monoclonal Antibody

£38.52 - £506.58

Specifications

Antigen TCR β chain
Clone H57-597
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15897048
View Documents
BD Biosciences
553168
0.1 mg
ONLINE EXCLUSIVE PRICE
£40.70
£38.52 / Each
Save £2.18
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15807058
View Documents
BD Biosciences
553169
0.5 mg
ONLINE EXCLUSIVE PRICE
£530.00
£506.58 / Each
Save £23.42
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The H57-597 antibody reacts with a common epitope of the β; chain of the T-cell Receptor (TCR) complex on α;β; TCR-expressing thymocytes and peripheral T lymphocytes and NK1.1+ thymocytes and NK-T cells of all mouse strains tested. It does not react with γδ TCR-bearing T cells. In the fetal and adult thymus, the TCR β; -chain may form homodimers or pair with the pre-TCR α; -chain on the surface of immature thymocytes before expression of the TCR α; -chain. Plate-bound or soluble H57-597 antibody activates α;β; TCR-bearing T cells, and plate-bound mAb can induce apoptotic death.
Specifications

Specifications

TCR β chain
0.5mg/mL
Monoclonal
Hamster
Immunology
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG2
Affinity Purified
H57-597
Flow Cytometry
Biotin
RUO
Mouse
TCR affinity-purified from mouse T-cell hybridoma DO-11.10
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.