missing translation for 'onlineSavingsMsg'
Learn More

TCR β chain Hamster anti-Mouse, Biotin, Clone: H57-597, BD

Hamster Monoclonal Antibody

Brand:  BD Biosciences 553168

 View more versions of this product

Product Code. 15897048

  • ONLINE EXCLUSIVE PRICE
    £40.70
    £38.52 / Each
    Save £2.18
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The H57-597 antibody reacts with a common epitope of the β; chain of the T-cell Receptor (TCR) complex on α;β; TCR-expressing thymocytes and peripheral T lymphocytes and NK1.1+ thymocytes and NK-T cells of all mouse strains tested. It does not react with γδ TCR-bearing T cells. In the fetal and adult thymus, the TCR β; -chain may form homodimers or pair with the pre-TCR α; -chain on the surface of immature thymocytes before expression of the TCR α; -chain. Plate-bound or soluble H57-597 antibody activates α;β; TCR-bearing T cells, and plate-bound mAb can induce apoptotic death.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TCR β chain
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
TCR affinity-purified from mouse T-cell hybridoma DO-11.10
0.1 mg
Immunology
Mouse
IgG2
Flow Cytometry
H57-597
Biotin
Hamster
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.