missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR β chain Hamster anti-Mouse, Biotin, Clone: H57-597, BD
Hamster Monoclonal Antibody
Brand: BD Biosciences 553169
This item is not returnable.
View return policy
Description
The H57-597 antibody reacts with a common epitope of the β; chain of the T-cell Receptor (TCR) complex on α;β; TCR-expressing thymocytes and peripheral T lymphocytes and NK1.1+ thymocytes and NK-T cells of all mouse strains tested. It does not react with γδ TCR-bearing T cells. In the fetal and adult thymus, the TCR β; -chain may form homodimers or pair with the pre-TCR α; -chain on the surface of immature thymocytes before expression of the TCR α; -chain. Plate-bound or soluble H57-597 antibody activates α;β; TCR-bearing T cells, and plate-bound mAb can induce apoptotic death.
Specifications
| TCR β chain | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| TCR affinity-purified from mouse T-cell hybridoma DO-11.10 | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG2 |
| Flow Cytometry | |
| H57-597 | |
| Biotin | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction