missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGN38 Antibody (2F7.1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
£205.00 - £482.00
Specifications
| Antigen | TGN38 |
|---|---|
| Clone | 2F7.1 |
| Dilution | Western Blot 1:500, Flow Cytometry 1:20 - 1:50, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18765643
|
Novus Biologicals
NB300-575 |
100 μL |
£482.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18788813
|
Novus Biologicals
NB300-575SS |
0.02 mL |
£205.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TGN38 Monoclonal specifically detects TGN38 in Human, Mouse, Rat, Chinese Hamster, Monkey, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TGN38 | |
| Western Blot 1:500, Flow Cytometry 1:20 - 1:50, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat, Chinese Hamster, Monkey, Rabbit | |
| 10618 | |
| Synthetic peptide corresponding to residues L(1) P S A S K P N N T S S E N N P P(17) C of rat TGN38. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 2F7.1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Neutralization | |
| Unconjugated | |
| Mouse | |
| Golgi Apparatus Markers, Trans golgi Network Markers | |
| TGN46TGN38 homolog, TGN48MGC14722, TGN5148, 51kD isoforms), trans-golgi network protein 2, Trans-Golgi network protein TGN51 | |
| TGOLN2 | |
| IgG1 | |
| Protein A or G purified | |
| Detects Trans Golgi Network (TGN) 38 from human and monkey samples and both recombinant and endogenous rat samples. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title