missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGN38 Antibody (2F7.1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
Brand: Novus Biologicals NB300-575SS
This item is not returnable.
View return policy
Description
TGN38 Monoclonal specifically detects TGN38 in Human, Mouse, Rat, Chinese Hamster, Monkey, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TGN38 | |
| Monoclonal | |
| Unconjugated | |
| TGN46TGN38 homolog, TGN48MGC14722, TGN5148, 51kD isoforms), trans-golgi network protein 2, Trans-Golgi network protein TGN51 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Detects Trans Golgi Network (TGN) 38 from human and monkey samples and both recombinant and endogenous rat samples. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Neutralization | |
| 2F7.1 | |
| Western Blot 1:500, Flow Cytometry 1:20 - 1:50, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000 | |
| TGOLN2 | |
| Synthetic peptide corresponding to residues L(1) P S A S K P N N T S S E N N P P(17) C of rat TGN38. | |
| 0.02 mL | |
| Golgi Apparatus Markers, Trans golgi Network Markers | |
| 10618 | |
| Human, Mouse, Rat, Chinese Hamster, Monkey, Rabbit | |
| Ascites |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido