missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGN38 Antibody (2F7.1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
Brand: Novus Biologicals NB300-575
This item is not returnable.
View return policy
Description
TGN38 Monoclonal specifically detects TGN38 in Human, Mouse, Rat, Chinese Hamster, Monkey, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TGN38 | |
| Monoclonal | |
| Unconjugated | |
| TGN46TGN38 homolog, TGN48MGC14722, TGN5148, 51kD isoforms), trans-golgi network protein 2, Trans-Golgi network protein TGN51 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Detects Trans Golgi Network (TGN) 38 from human and monkey samples and both recombinant and endogenous rat samples. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Neutralization | |
| 2F7.1 | |
| Western Blot 1:500, Flow Cytometry 1:20 - 1:50, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000 | |
| TGOLN2 | |
| Synthetic peptide corresponding to residues L(1) P S A S K P N N T S S E N N P P(17) C of rat TGN38. | |
| 100 μL | |
| Golgi Apparatus Markers, Trans golgi Network Markers | |
| 10618 | |
| Human, Mouse, Rat, Chinese Hamster, Monkey, Rabbit | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction