Learn More
GSK-3β Mouse, Unlabeled, Clone: 7, BD
Mouse Monoclonal Antibody
£157.25 - £401.97
Specifications
| Antigen | GSK-3β |
|---|---|
| Clone | 7 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15855329
|
BD Biosciences
610201 |
50 μg |
£164.00 £157.25 / 1 set Save £6.75 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15371687
|
BD Biosciences
610202 |
150 μg |
£419.00 £401.97 / Each Save £17.03 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycogen Synthase Kinase-3β (GSK-3β) is a serine/threonine kinase that affects glycogen metabolism by phosphorylating and, thus, down-regulating the activity of muscle glycogen synthase. GSK-3β is identical to the Tau Protein Kinase 1 (TPK 1) which plays a role in the formation of the histopathological lesions of Alzheimer's disease (AD). In AD, many neurons contain intracytoplasmic neurofibrillary tangles (NFT) that are composed of ubiquitin and highly phosphorylated tau proteins, members of a family of microtubule-assocaited phosphoproteins. Phosphorylation of the tau proteins by GSK-3β converts them into paired helical filaments (PHF) which are found in NFT and degenerative neurites of senile plaques. This ability of GSK-3β to convert tau proteins to PHF has been demonstrated in vitro using primary cultures of embryonic rat hippocampal neurons.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| GSK-3β | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 7 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Chicken, Human, Mouse, Rat | |
| Rat GSK-3β aa. 1-160 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.