missing translation for 'onlineSavingsMsg'
Learn More

GSK-3β Mouse, Unlabeled, Clone: 7, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610201

 View more versions of this product

Product Code. 15855329

  • ONLINE EXCLUSIVE PRICE
    £164.00
    £157.25 / 1 set
    Save £6.75
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Glycogen Synthase Kinase-3β (GSK-3β) is a serine/threonine kinase that affects glycogen metabolism by phosphorylating and, thus, down-regulating the activity of muscle glycogen synthase. GSK-3β is identical to the Tau Protein Kinase 1 (TPK 1) which plays a role in the formation of the histopathological lesions of Alzheimer's disease (AD). In AD, many neurons contain intracytoplasmic neurofibrillary tangles (NFT) that are composed of ubiquitin and highly phosphorylated tau proteins, members of a family of microtubule-assocaited phosphoproteins. Phosphorylation of the tau proteins by GSK-3β converts them into paired helical filaments (PHF) which are found in NFT and degenerative neurites of senile plaques. This ability of GSK-3β to convert tau proteins to PHF has been demonstrated in vitro using primary cultures of embryonic rat hippocampal neurons.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

GSK-3β
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Rat GSK-3β aa. 1-160
50 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG1
Western Blot
7
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.