Learn More
GSK-3β Mouse, Unlabeled, Clone: 7, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610202
Description
Glycogen Synthase Kinase-3β (GSK-3β) is a serine/threonine kinase that affects glycogen metabolism by phosphorylating and, thus, down-regulating the activity of muscle glycogen synthase. GSK-3β is identical to the Tau Protein Kinase 1 (TPK 1) which plays a role in the formation of the histopathological lesions of Alzheimer's disease (AD). In AD, many neurons contain intracytoplasmic neurofibrillary tangles (NFT) that are composed of ubiquitin and highly phosphorylated tau proteins, members of a family of microtubule-assocaited phosphoproteins. Phosphorylation of the tau proteins by GSK-3β converts them into paired helical filaments (PHF) which are found in NFT and degenerative neurites of senile plaques. This ability of GSK-3β to convert tau proteins to PHF has been demonstrated in vitro using primary cultures of embryonic rat hippocampal neurons.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| GSK-3β | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Rat GSK-3β aa. 1-160 | |
| 150 μg | |
| Cell Biology | |
| Canine, Chicken, Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| 7 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.