Learn More
CD45 Mouse anti-Human, APC-H7, Clone: 2D1, BD
Mouse Monoclonal Antibody
£127.91 - £422.47
Specifications
| Antigen | CD45 |
|---|---|
| Clone | 2D1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC-H7 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15840069
|
BD Biosciences
560274 |
25 Tests |
£133.00 £127.91 / Each Save £5.09 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15869988
|
BD Biosciences
560178 |
100 Tests |
£428.00 £422.47 / Each Save £5.53 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The clone 2D1 recognizes the CD45 antigen, a tyrosine phosphatase. There are several isoforms of CD45 with molecular weights ranging from 180 to 220kDa and all are members of the T200 family. The CD45 antigen is present on all human leukocytes, including lymphocytes, monocytes, granulocytes, eosinophils and basophils in peripheral blood and has a role in signal transduction modifying signals from other surface molecules.
Host Species: Mouse
Clone: 2D1
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD45 | |
| Flow Cytometry | |
| APC-H7 | |
| RUO | |
| Human | |
| IgG1 κ | |
| Affinity Purified | |
| Leukocyte Common Antigen |
| 2D1 | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.