missing translation for 'onlineSavingsMsg'
Learn More

CD45 Mouse anti-Human, APC-H7, Clone: 2D1, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 560178

 View more versions of this product

Product Code. 15869988

  • ONLINE EXCLUSIVE PRICE
    £428.00
    £422.47 / Each
    Save £5.53
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The clone 2D1 recognizes the CD45 antigen, a tyrosine phosphatase. There are several isoforms of CD45 with molecular weights ranging from 180 to 220kDa and all are members of the T200 family. The CD45 antigen is present on all human leukocytes, including lymphocytes, monocytes, granulocytes, eosinophils and basophils in peripheral blood and has a role in signal transduction modifying signals from other surface molecules.

Host Species: Mouse
Clone: 2D1
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD45
Monoclonal
APC-H7
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
2D1
Leukocyte Common Antigen
Mouse
100 Tests
Immunology
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.