missing translation for 'onlineSavingsMsg'
Learn More

CD45 Mouse anti-Human, APC-H7, Clone: 2D1, BD

Product Code. 15840069
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15840069 25 Tests Each
15869988 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15840069 Supplier BD Biosciences Supplier No. 560274

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The clone 2D1 recognizes the CD45 antigen, a tyrosine phosphatase. There are several isoforms of CD45 with molecular weights ranging from 180 to 220kDa and all are members of the T200 family. The CD45 antigen is present on all human leukocytes, including lymphocytes, monocytes, granulocytes, eosinophils and basophils in peripheral blood and has a role in signal transduction modifying signals from other surface molecules.

Host Species: Mouse
Clone: 2D1
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45
Applications Flow Cytometry
Classification Monoclonal
Clone 2D1
Conjugate APC-H7
Description Leukocyte Common Antigen
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.