Learn More
Annexin II (Lipocortin II, Calpactin I) Mouse anti-Human, FITC, Clone: 5, BD
Mouse Monoclonal Antibody
£353.00 - £540.33
Specifications
| Antigen | Annexin II (Lipocortin II, Calpactin I) |
|---|---|
| Clone | 5 |
| Concentration | 250μg/mL |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
Description
Annexin II (p36), originally discovered as a substrate for the src oncogene, is one of the most studied members of the annexin family of calcium-dependent phospholipid-binding proteins. It exists in cells as either a monomer or a heterotetramer complexed with p11, an S100-related protein. In vitro, it has been found that unphosphorylated annexin II is capable of causing aggregation of chromaffin granules, and, upon phosphorylation, induces fusion of the granules' membranes. A similiar phenomenon may occur in the final steps of exocytosis after cytoplasmic annexin translocates to the subplasmalemmal region and is phosphorylated by protein kinase C. However, other studies refute annexin II's involvement in exocytosis, and a conclusive role for annexin II in membrane trafficking remains debated.
Host Species: Mouse
Clone: 5
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human Annein II aa. 123-328
Immunofluorescence, Immunoprecipitation
Specifications
| Annexin II (Lipocortin II, Calpactin I) | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified | |
| Lipocortin II, Calpactin I |
| 5 | |
| Immunofluorescence | |
| FITC | |
| RUO | |
| Human | |
| Human Annein II aa. 123-328 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.