Learn More
Annexin II (Lipocortin II, Calpactin I) Mouse anti-Human, FITC, Clone: 5, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 611839
Description
Annexin II (p36), originally discovered as a substrate for the src oncogene, is one of the most studied members of the annexin family of calcium-dependent phospholipid-binding proteins. It exists in cells as either a monomer or a heterotetramer complexed with p11, an S100-related protein. In vitro, it has been found that unphosphorylated annexin II is capable of causing aggregation of chromaffin granules, and, upon phosphorylation, induces fusion of the granules' membranes. A similiar phenomenon may occur in the final steps of exocytosis after cytoplasmic annexin translocates to the subplasmalemmal region and is phosphorylated by protein kinase C. However, other studies refute annexin II's involvement in exocytosis, and a conclusive role for annexin II in membrane trafficking remains debated.
Host Species: Mouse
Clone: 5
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human Annein II aa. 123-328
Immunofluorescence, Immunoprecipitation
Specifications
| Annexin II (Lipocortin II, Calpactin I) | |
| Monoclonal | |
| 250μg/mL | |
| Lipocortin II, Calpactin I | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Immunofluorescence | |
| 5 | |
| FITC | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Human Annein II aa. 123-328 | |
| 150 μg | |
| Cell Biology | |
| Human | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.