missing translation for 'onlineSavingsMsg'
Learn More

Annexin II (Lipocortin II, Calpactin I) Mouse anti-Human, FITC, Clone: 5, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611838

 View more versions of this product

Product Code. 15836039

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Annexin II (p36), originally discovered as a substrate for the src oncogene, is one of the most studied members of the annexin family of calcium-dependent phospholipid-binding proteins. It exists in cells as either a monomer or a heterotetramer complexed with p11, an S100-related protein. In vitro, it has been found that unphosphorylated annexin II is capable of causing aggregation of chromaffin granules, and, upon phosphorylation, induces fusion of the granules' membranes. A similiar phenomenon may occur in the final steps of exocytosis after cytoplasmic annexin translocates to the subplasmalemmal region and is phosphorylated by protein kinase C. However, other studies refute annexin II's involvement in exocytosis, and a conclusive role for annexin II in membrane trafficking remains debated.

Host Species: Mouse
Clone: 5
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human Annein II aa. 123-328

Immunofluorescence, Immunoprecipitation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Annexin II (Lipocortin II, Calpactin I)
Monoclonal
250μg/mL
Lipocortin II, Calpactin I
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Immunofluorescence
5
FITC
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human Annein II aa. 123-328
50 μg
Cell Biology
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.