missing translation for 'onlineSavingsMsg'
Learn More

Annexin II Light Chain Mouse, Unlabeled, Clone: 148, BD

Mouse Monoclonal Antibody

£223.06 - £284.36

Specifications

Antigen Annexin II Light Chain
Clone 148
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15855239
View Documents
BD Biosciences
610070
50 μg
ONLINE EXCLUSIVE PRICE
£233.00
£223.06 / 1 set
Save £9.94
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15865239
View Documents
BD Biosciences
610071
150 μg
ONLINE EXCLUSIVE PRICE
£297.00
£284.36 / 1 set
Save £12.64
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Annexin II often exists as a heterotetramer complexed with an 11kDa light chain. This p11 subunit is related to the S100 family of calcium-binding proteins but mutations have rendered it unable to bind calcium. The p11 protein binds to the amino terminus of the annexin II monomer, but not to other annexins. It may regulate the function of annexin II, as the tetrameric form seems to have a higher calcium and phospholipid-binding affinity than the monomeric annexin II. In the cell, p11 has been localized to the cytoplasmic face of the plasma membrane where the annexin II tetramer is known to exist.

Host Species: Mouse
Clone: 148
Isotype: IgG1
Species Reactivity: Human
Immunogen: Cow Annexin II Light Chain aa. 1-97
Formula Weight [Chemical]: 11kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Annexin II Light Chain
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
148
Western Blot
Unconjugated
RUO
Canine, Chicken, Human
Cow Annexin II Light Chain aa. 1-97
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.