Learn More
Annexin II Light Chain Mouse, Unlabeled, Clone: 148, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610070
Description
Annexin II often exists as a heterotetramer complexed with an 11kDa light chain. This p11 subunit is related to the S100 family of calcium-binding proteins but mutations have rendered it unable to bind calcium. The p11 protein binds to the amino terminus of the annexin II monomer, but not to other annexins. It may regulate the function of annexin II, as the tetrameric form seems to have a higher calcium and phospholipid-binding affinity than the monomeric annexin II. In the cell, p11 has been localized to the cytoplasmic face of the plasma membrane where the annexin II tetramer is known to exist.
Host Species: Mouse
Clone: 148
Isotype: IgG1
Species Reactivity: Human
Immunogen: Cow Annexin II Light Chain aa. 1-97
Formula Weight [Chemical]: 11kDa
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| Annexin II Light Chain | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Cow Annexin II Light Chain aa. 1-97 | |
| 50 μg | |
| Cell Biology | |
| Canine, Chicken, Human | |
| IgG1 |
| Western Blot | |
| 148 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.