missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD44 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | ANKRD44 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18646222
|
Novus Biologicals
NBP2-92554-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18622102
|
Novus Biologicals
NBP2-92554-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ANKRD44 Polyclonal antibody specifically detects ANKRD44 in Human samples. It is validated for Western BlotSpecifications
| ANKRD44 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ankyrin repeat domain 44, FLJ11570, MGC21968, MGC70444, PP6-ARS-BAnkyrin repeat domain-containing protein 44, protein phosphatase 6 ankyrin repeat subunit B, serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit B, Serine/threonine-protein phosphatase 6 regulatory subunit ARS-B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 310-550 of human ANKRD44 (NP_001182073.1). LHMTAVHGRFTRSQTLIQNGGEIDCVDKDGNTPLHVAARYGHELLINTLITSGADTAKCGIHSMFPLHLAALNAHSDCCRKLLSSGFEIDTPDKFGRTCLHAAAAGGNVECIKLLQSSGADFHKKDKCGRTPLHYAAANCHFHCIETLVTTGANVNETDDWGRTALHYAAASDMDRNKTILGNAHDNSEELERARELKEKEATLCLEFLLQNDANPSIRDKEGYNSIHYAAAYGHRQCLEL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 91526 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title