missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD44 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92554-0.1ml
This item is not returnable.
View return policy
Description
ANKRD44 Polyclonal antibody specifically detects ANKRD44 in Human samples. It is validated for Western Blot
Specifications
| ANKRD44 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ankyrin repeat domain 44, FLJ11570, MGC21968, MGC70444, PP6-ARS-BAnkyrin repeat domain-containing protein 44, protein phosphatase 6 ankyrin repeat subunit B, serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit B, Serine/threonine-protein phosphatase 6 regulatory subunit ARS-B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 310-550 of human ANKRD44 (NP_001182073.1). LHMTAVHGRFTRSQTLIQNGGEIDCVDKDGNTPLHVAARYGHELLINTLITSGADTAKCGIHSMFPLHLAALNAHSDCCRKLLSSGFEIDTPDKFGRTCLHAAAAGGNVECIKLLQSSGADFHKKDKCGRTPLHYAAANCHFHCIETLVTTGANVNETDDWGRTALHYAAASDMDRNKTILGNAHDNSEELERARELKEKEATLCLEFLLQNDANPSIRDKEGYNSIHYAAAYGHRQCLEL | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 91526 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction