missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD44 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92554-0.02ml
This item is not returnable.
View return policy
Description
ANKRD44 Polyclonal antibody specifically detects ANKRD44 in Human samples. It is validated for Western Blot
Specifications
| ANKRD44 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ankyrin repeat domain 44, FLJ11570, MGC21968, MGC70444, PP6-ARS-BAnkyrin repeat domain-containing protein 44, protein phosphatase 6 ankyrin repeat subunit B, serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit B, Serine/threonine-protein phosphatase 6 regulatory subunit ARS-B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 310-550 of human ANKRD44 (NP_001182073.1). LHMTAVHGRFTRSQTLIQNGGEIDCVDKDGNTPLHVAARYGHELLINTLITSGADTAKCGIHSMFPLHLAALNAHSDCCRKLLSSGFEIDTPDKFGRTCLHAAAAGGNVECIKLLQSSGADFHKKDKCGRTPLHYAAANCHFHCIETLVTTGANVNETDDWGRTALHYAAASDMDRNKTILGNAHDNSEELERARELKEKEATLCLEFLLQNDANPSIRDKEGYNSIHYAAAYGHRQCLEL | |
| 0.02 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 91526 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion