missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ALK, GST Tag Recombinant Protein
Anaplastic Lymphoma Receptor Tyrosine Kinase (ALK) belongs to the insulin receptor superfamily.
£437.00 - £6530.00
Specifications
Gene Alias | CD246, NBLST3 |
---|---|
KinaseFamily | ALK Kinase Family |
Species | Human |
Recombinant | Recombinant |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11543597
|
Invitrogen™
PV3867 |
10μg |
£437.00
10µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15418583
|
Invitrogen™
PR7396B |
100μg |
£1460.00
100µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10188492
|
Invitrogen™
PV4185 |
1mg |
£6530.00
1mg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ALK is vital for brain development. Mutations, rearrangements, and amplifications in the ALK gene have been found in tumors, including anaplastic large cell lymphoma, neurblastoma, and non-small cell lung cancer. Chromosomal rearrangement is the most common genetic alteration.
Specifications
CD246, NBLST3 | |
Human | |
Baculovirus |
ALK Kinase Family | |
Recombinant |