Learn More
Invitrogen™ Human ALK, GST Tag Recombinant Protein
Anaplastic Lymphoma Receptor Tyrosine Kinase (ALK) belongs to the insulin receptor superfamily.
Brand: Invitrogen™ PV3867
Code : NEW
Additional Details : CAS Number : 9048-46-8 Weight : 0.00900kg
Description
ALK is vital for brain development. Mutations, rearrangements, and amplifications in the ALK gene have been found in tumors, including anaplastic large cell lymphoma, neurblastoma, and non-small cell lung cancer. Chromosomal rearrangement is the most common genetic alteration.
Specifications
NP_004295, NP_004295.2, J1461V | |
114294, 238, 238 | |
10μg | |
ALK Kinase Family | |
Human | |
Kinases & Inhibitors | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
ALK | |
Baculovirus | |
Recombinant, Active, Cytoplasmic | |
Kinases & Inhibitors |
Kinase Assay | |
89.9 kDa, 90 kDa | |
CD246, NBLST3 | |
aa 1058-1620 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
anaplastic lymphoma receptor tyrosine kinase | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |