Learn More
Invitrogen™ Human ALK, GST Tag Recombinant Protein
Anaplastic Lymphoma Receptor Tyrosine Kinase (ALK) belongs to the insulin receptor superfamily.
Brand: Invitrogen™ PR7396B
Code : NEW
Additional Details : Weight : 0.01000kg
Description
ALK is vital for brain development. Mutations, rearrangements, and amplifications in the ALK gene have been found in tumors, including anaplastic large cell lymphoma, neurblastoma, and non-small cell lung cancer. Chromosomal rearrangement is the most common genetic alteration.
Specifications
NP_004295 | |
238 | |
100μg | |
ALK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
|
Baculovirus | |
Recombinant, Active, Cytoplasmic | |
Receptor Tyrosine Kinases |
Kinase Assay | |
89.9 kDa | |
CD246, NBLST3 | |
Human | |
GST Tag | |
Receptor Tyrosine Kinase | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |