missing translation for 'onlineSavingsMsg'
Learn More

Adenine Nucleotide Translocase 1 Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody

£154.00 - £368.00

Specifications

Antigen Adenine Nucleotide Translocase 1
Dilution Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30232620
View Documents
Novus Biologicals
NBP3-33484-100ul
100 μL
£368.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30227900
View Documents
Novus Biologicals
NBP3-33484-20ul
20 μL
£154.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Adenine Nucleotide Translocase 1 Monoclonal antibody specifically detects Adenine Nucleotide Translocase 1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications

Specifications

Adenine Nucleotide Translocase 1
ELISA, Western Blot
Unconjugated
Rabbit
Apoptosis, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
PBS (pH 7.3), 50% glycerol, 0.05% BSA
291
IgG
Affinity purified
Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL
Monoclonal
Purified
RUO
Human, Mouse, Rat
AAC1, Adenine nucleotide translocator 1, ADP, ADP/ATP translocase 1, ANT 1, ANT1adenine nucleotide translocator 1 (skeletal muscle), ATP carrier protein 1, ATP carrier protein, heart/skeletal muscle, ATP carrier protein, heart/skeletal muscle isoform T1, heart/skeletal muscle ATP/ADP translocator, PEO2, PEO3, solute carrier family 25 (mitochondrial carrier; adenine nucleotidetranslocator), member 4, Solute carrier family 25 member 4, T1ANT
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Adenine Nucleotide Translocase 1 (NP_001142.2).,, Sequence:, MGDHAWSFLKDFLAGGVAAAVSKTAVAPIERVKLLLQVQHASKQISAEKQYKGIIDCVVRIPKEQGFLSFWRGNLANVIRYFPTQALNFAFKDKYKQLFLGGVDRHKQFWRYFAGNLASGGAAGATSLCFVYPLDFARTRLAADVGKGAAQREFHGLGDCIIKIFKSDGLRGLYQGFNVSVQGIIIYRAAYFGVYDTAKG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.