missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenine Nucleotide Translocase 1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33484-20ul
This item is not returnable.
View return policy
Description
Adenine Nucleotide Translocase 1 Monoclonal antibody specifically detects Adenine Nucleotide Translocase 1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Adenine Nucleotide Translocase 1 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| AAC1, Adenine nucleotide translocator 1, ADP, ADP/ATP translocase 1, ANT 1, ANT1adenine nucleotide translocator 1 (skeletal muscle), ATP carrier protein 1, ATP carrier protein, heart/skeletal muscle, ATP carrier protein, heart/skeletal muscle isoform T1, heart/skeletal muscle ATP/ADP translocator, PEO2, PEO3, solute carrier family 25 (mitochondrial carrier; adenine nucleotidetranslocator), member 4, Solute carrier family 25 member 4, T1ANT | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Adenine Nucleotide Translocase 1 (NP_001142.2).,, Sequence:, MGDHAWSFLKDFLAGGVAAAVSKTAVAPIERVKLLLQVQHASKQISAEKQYKGIIDCVVRIPKEQGFLSFWRGNLANVIRYFPTQALNFAFKDKYKQLFLGGVDRHKQFWRYFAGNLASGGAAGATSLCFVYPLDFARTRLAADVGKGAAQREFHGLGDCIIKIFKSDGLRGLYQGFNVSVQGIIIYRAAYFGVYDTAKG | |
| 20 μL | |
| Apoptosis, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 291 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction