missing translation for 'onlineSavingsMsg'
Learn More

Adenine Nucleotide Translocase 1 Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-33484-100ul

 View more versions of this product

Product Code. 30232620

  • £368.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Adenine Nucleotide Translocase 1 Monoclonal antibody specifically detects Adenine Nucleotide Translocase 1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Adenine Nucleotide Translocase 1
Monoclonal
Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL
AAC1, Adenine nucleotide translocator 1, ADP, ADP/ATP translocase 1, ANT 1, ANT1adenine nucleotide translocator 1 (skeletal muscle), ATP carrier protein 1, ATP carrier protein, heart/skeletal muscle, ATP carrier protein, heart/skeletal muscle isoform T1, heart/skeletal muscle ATP/ADP translocator, PEO2, PEO3, solute carrier family 25 (mitochondrial carrier; adenine nucleotidetranslocator), member 4, Solute carrier family 25 member 4, T1ANT
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Adenine Nucleotide Translocase 1 (NP_001142.2).,, Sequence:, MGDHAWSFLKDFLAGGVAAAVSKTAVAPIERVKLLLQVQHASKQISAEKQYKGIIDCVVRIPKEQGFLSFWRGNLANVIRYFPTQALNFAFKDKYKQLFLGGVDRHKQFWRYFAGNLASGGAAGATSLCFVYPLDFARTRLAADVGKGAAQREFHGLGDCIIKIFKSDGLRGLYQGFNVSVQGIIIYRAAYFGVYDTAKG
100 μL
Apoptosis, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
291
Store at -20°C. Avoid freeze-thaw cycles.
IgG
ELISA, Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.