missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated Charcoal, Tested according to Ph.Eur., Honeywell™ Fluka™
tested according to Ph. Eur.
£460.38 - £2603.05
Chemical Identifiers
CAS | 7440-44-0 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12.011 |
MDL Number | MFCD00133992 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
PubChem CID | 5462310 |
ChEBI | CHEBI:27594 |
IUPAC Name | carbon |
SMILES | [C] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15630790
|
Honeywell-Fluka
05117-1KG |
1 kg | Plastic Bottle | ||||||
15620790
|
Honeywell-Fluka
05117-10KG |
10 kg | Plastic Drum |
£2603.05
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7440-44-0 | |
12.011 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
C | |
MFCD00133992 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
Specifications
7440-44-0 | |
Black | |
6 to 9 | |
MFCD00133992 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
[C] | |
12.011 | |
CHEBI:27594 | |
Activated Charcoal, Tested According to Ph.Eur. |
3550°C (literature) | |
ca. 350 kg/cu.m | |
C | |
NONH for all modes of transport | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
12.01g/mol |
Safety and Handling
missing translation for 'einecsNumber' : 231-153-3