missing translation for 'onlineSavingsMsg'
Learn More

Activated Charcoal, Tested according to Ph.Eur., Honeywell™ Fluka™

Product Code. 15620790 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
10kg
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15620790 10 kg Plastic Drum 10kg
15630790 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15620790 Supplier Honeywell-Fluka Supplier No. 0511710KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

tested according to Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Melting Point 3550°C (literature)
Color Black
Density ca. 350 kg/cu.m
pH 6 to 9
Packaging Plastic Drum
Quantity 10 kg
UN Number NONH for all modes of transport
Formula Weight 12.01g/mol
Chemical Name or Material Activated Charcoal, Tested According to Ph.Eur.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.