Learn More
Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with Nylon Membrane
Versatile filter for both aqueous and solvent-based sample filtration.
£126.00 - £2695.00
Specifications
Sterility | Non-sterile |
---|---|
Filtration Area | 3.9 cm2 |
Housing Material | Polypropylene |
Product Code | Brand | Pore Size | No. per Pack | Material (Membrane) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | No. per Pack | Material (Membrane) | Price | Quantity & Availability | |||
17184671
|
Cytiva (formerly Pall Lab)
AP-4438 |
0.45 μm | 50/Pk. | Nylon |
£126.00
Pack of 50 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
17134681
|
Cytiva (formerly Pall Lab)
AP-4522 |
0.2 μm | 1000/Pk. | Nylon |
£2255.00
Pack of 1000 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
17174671
|
Cytiva (formerly Pall Lab)
AP-4436 |
0.2 μm | 50/Pk. | Nylon |
£126.00
Pack of 50 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||
17184681
|
Cytiva (formerly Pall Lab)
AP-4549 |
0.45 μm | 50/Pk. | Glass Fiber, Nylon |
£150.00
Pack of 50 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
17114681
|
Cytiva (formerly Pall Lab)
AP-4502 |
0.45 μm | 1000/Pk. | Nylon |
£2695.00
Pack of 1000 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||
17154681
|
Cytiva (formerly Pall Lab)
AP-4528 |
0.45 μm | 1000/Pk. | Glass Fiber, Nylon |
£2255.00
Pack of 1000 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||
17134691
|
Cytiva (formerly Pall Lab)
AP-4787 |
0.2 μm | 50/Pk. | Glass Fiber, Nylon |
£149.00
Pack of 50 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
17144691
|
Cytiva (formerly Pall Lab)
AP-4788 |
0.2 μm | 1000/Pk. | Glass Fiber, Nylon |
£2255.00
Pack of 1000 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
17174681
|
Cytiva (formerly Pall Lab)
AP-4548 |
0.45 μm | 25/AutoPack Tubes | Glass Fiber, Nylon |
£640.00
Pack of 200 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
17124691
|
Cytiva (formerly Pall Lab)
AP-4786 |
0.2 μm | 25/AutoPack Tubes | Glass Fiber, Nylon |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Excellent chemical compatibility with esters, bases, and alcohols
- Saves time because no prewetting is required
Specifications
Non-sterile | |
3.9 cm2 | |
Polypropylene |