missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with Nylon Membrane

Versatile filter for both aqueous and solvent-based sample filtration.

Brand:  Cytiva (formerly Pall Lab) AP-4438

109.29 GBP valid until 2024-05-06
Use promo code "21711" to get your promotional price.



Additional Details : Weight : 0.98800kg

 View more versions of this product

Product Code. 17184671

  • £126.00 / Pack of 50
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

  • Excellent chemical compatibility with esters, bases, and alcohols
  • Saves time because no prewetting is required
Specifications

Specifications

Nylon
Non-sterile
Polypropylene
25 mm
0.45 μm
3.9 cm2
50/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers