missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with Nylon Membrane
Versatile filter for both aqueous and solvent-based sample filtration.
Brand: Cytiva (formerly Pall Lab) AP-4502
2560.25 GBP valid until 2024-05-06
Use promo code "21711" to get your promotional price.
Additional Details : Weight : 1.81450kg
Description
- Excellent chemical compatibility with esters, bases, and alcohols
- Saves time because no prewetting is required
Specifications
Nylon | |
Non-sterile | |
Polypropylene | |
25 mm |
0.45 μm | |
3.9 cm2 | |
1000/Pk. |