Learn More
Waters Corporation ACQUITY UPLC BEH Phenyl Column
ACQUITY BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
£1042.50 - £2910.00
Specifications
Max. Pressure | 18000 psi (1240 Bar) |
---|---|
Particle Shape | Spherical |
Particle Size | 1.7 μm |
Pore Size | 130 Å |
USP Type | L11 |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Quantity | Price | Quantity & Availability | |||
12603456
|
Waters Corporation
186002884 |
2.1 mm | 50 mm | 1/Ea. |
£1072.50
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||
10710347
|
Waters Corporation
186002885 |
2.1 mm | 100 mm | 1/Ea. |
£1132.50
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||
13135069
|
Waters Corporation
186003378 |
2.1 mm | 150 mm | 1/Ea. |
£1215.00
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||
13105069
|
Waters Corporation
186003375 |
1 mm | 150 mm | 1/Ea. |
£1215.00
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||
13283250
|
Waters Corporation
186003911 |
2.1 mm | 30 mm | 1/Ea. |
£1042.50
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||
13199939
|
Waters Corporation
176001311 |
2.1 mm | 30 mm | 3/Pk. |
£2910.00
Pack of 3 |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||
10135873
|
Waters Corporation
186002882 |
1 mm | 50 mm | 1/Ea. |
£1072.50
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||
13222920
|
Waters Corporation
186002883 |
1 mm | 100 mm | 1/Ea. |
£1132.50
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||
10459014
|
Waters Corporation
186004672 |
3 mm | 50 mm | 1/Ea. |
£1072.50
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||
13233820
|
Waters Corporation
186004673 |
3 mm | 100 mm | 1/Ea. |
£1132.50
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||
13223820
|
Waters Corporation
186004671 |
3 mm | 30 mm | 1/Ea. |
£1042.50
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||
13243820
|
Waters Corporation
186004674 |
3 mm | 150 mm | 1/Ea. |
£1215.00
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||
17103212
|
Waters Corporation
186005607 |
2.1 mm | 75 mm | 1/Ea. |
£1110.00
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||
17153212
|
Waters Corporation
186005612 |
3 mm | 75 mm | 1/Ea. |
£1110.00
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||
Description
- This sorbent, along with the XBridge BEH Phenyl, provide you with the most chemically stable phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape.
Specifications
18000 psi (1240 Bar) | |
1.7 μm | |
L11 | |
Column |
Spherical | |
130 Å | |
Yes |