missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AR430 Conductivity Meter with Stand

Product Code. 31702807 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Type:
Conductivity Meter
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
31702807 Conductivity Meter 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31702807
  • ONLINE EXCLUSIVE PRICE
    £1490.00
    £805.37 / 1 sets
    Save £684.63
    45% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to deliver precise and reliable electrochemistry testing, the accumet AR430 offers enhanced performance in a streamlined design. Ideal for laboratories requiring dependable measurement capabilities.

  • Large 7-in. high-resolution color display with capacitive touchscreen operable with gloves.
  • One to five point conductivity calibration with automatic buffer/standard recognition.
  • Streamlined, easy-to-operate user interface with step-by-step calibration guidance.
  • Stores 10 calibration logs per parameter with date and time.
  • Stores 5000 data log sets with date and time.
  • Outputs to a printer, computer, or USB flash drive for long-term data retention.
  • Inputs sample ID with optional USB barcode scanner, USB keyboard, or USB mouse.
  • Stirs samples with meter-controlled and meter-powered stirrer probe.
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (Conductivity) ±1% reading >3 μS/cm; ±1% reading ±0.01 μS/cm ≤3 μS/cm
Measurement Modes Conductivity, TDS, Salinity, Resistivity, Temperature
Calibration Conductivity, TDS, Salinity, Temperature
Calibration Points 1
Range (Conductivity) 0.010 μS/cm to 3000 mS/cm
Resolution (Conductivity) 0.001 μS/cm minimum, auto-ranging, up to 4 digits
Range (Salinity) 0.010 to 80.00 ppt
Resolution (Salinity) 0.001 ppt minimum, auto-ranging
Accuracy (Salinity) ±1% reading
Range (TDS) 0.010 ppm to 500.0 ppt (TDS factor of 1.00)
Resolution (TDS) 0.001 ppm minimum, auto-ranging, up to 4 digits
Accuracy (TDS) ±1% reading
Temperature Range (Metric) -30.0 to 130.0°C
Resolution (Temperature) 0.1°C, 0.1°F
Accuracy (Temperature) ±0.3°C, ±0.5°F
Range (Resistivity) 1.0 Ω to 100.0 MΩ
Display Type 7-in. High-resolution Color Graphic LCD
Data Points 5000 with Date and Time Stamp
Log Function Type Automatic Data Logging
Certifications/Compliance CE, TUV 3-1, FCC Class A
IP Rating IP54
Resolution (Resistivity) 0.1 Ω minimum, auto-ranging
Channels 1
Type Conductivity Meter
Accuracy (Resistivity) ±1% reading
Voltage 100/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.