missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs 50 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
£136.00 - £155.00
Specifications
MWCO | 50 kDa |
---|
Includes:
- 1m Biotech CE Tubing in 0.05% sodium azide
- Two universal tubing closures
- Five opening picks
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
15380672
|
Spectrum™
131381T |
1.8 mL/cm vol./length | 15 mm |
£152.00
1m |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15370672
|
Spectrum™
131372T |
0.32 mL/cm vol./length | 6.4 mm |
£136.00
1m |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15390672
|
Spectrum™
131384T |
3.1 mL/cm vol./length | 20 mm |
£155.00
1m |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11540970
|
Spectrum™
131378T |
0.79 mL/cm vol./length | 10 mm |
£148.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
- Hydrophilic, symmetric porositiy
- For small tubing demand applications or evaluating a specific dialysis membrane
- Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifications
50 kDa |