missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs 50 kD Biotech CE Dialysis Kits
Click to view available options
Diameter (Metric):
6.4 mm
10 mm
15 mm
20 mm
Length (Metric):
0.32 mL/cm vol./length
0.79 mL/cm vol./length
1.8 mL/cm vol./length
3.1 mL/cm vol./length
Unit Size:
1m
Each
Description
Includes
- 1m Biotech CE Tubing in 0.05% sodium azide
- Two universal tubing closures
- Five opening picks
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
- Hydrophilic, symmetric porositiy
- For small tubing demand applications or evaluating a specific dialysis membrane
- Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifications
Specifications
| MWCO | 50 kDa |
| Diameter (Metric) | 6.4 mm |
| Length (Metric) | 0.32 mL/cm vol./length |
| Quantity | 1 m Tubing/Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction