missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs 50 kD Biotech CE Dialysis Kits

With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane

Brand:  Spectrum™ 131378T

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11540970

  • £148.00 / Each
Estimated Shipment: 26-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes:
  • 1m Biotech CE Tubing in 0.05% sodium azide
  • Two universal tubing closures
  • Five opening picks

Description

Description

Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC

  • Hydrophilic, symmetric porositiy
  • For small tubing demand applications or evaluating a specific dialysis membrane
  • Includes 1m tubing with pair of tubing closures and 5 opening picks

Specifications

Specifications

Dialysis Trial Kit
Cellulose Ester
0.39 in.
1m roll Biotech grade CE dialysis membrane tubing, 2 universal closures, 5 opening picks
Ideal for small, critical dialysis applications
50 kDa
10 mm
0.79 mL/cm vol./length
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers