missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 2-(Pyridyldithio)ethylamine hydrochloride

Product Code. p-200104411
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16470936

Brand: MedChemExpress HY101794

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 2-(Pyridyldithio)ethylamine hydrochloride is a novel disulfide intercalating cross-linking reagent.

Specifications

Molecular Weight (g/mol) 222.76 g/mol
CAS 106139-15-5
Content And Storage 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture)
Shipping Condition Room temperature
Quantity 200 mg
Synonym (S)-2-Pyridylthio Cysteamine Hydrochloride
Solubility H2O : 50 mg/mL (224.46 mM; Need ultrasonic) ∣ DMSO : ≥ 32 mg/mL (143.65 mM)
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.