missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 2-(Pyridyldithio)ethylamine hydrochloride

Product Code. 16470936
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470936 200 mg 200mg
16459986 50 mg 50mg
16451536 500 mg 500mg
16444686 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16470936 Supplier MedChemExpress Supplier No. HY101794

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 2-(Pyridyldithio)ethylamine hydrochloride is a novel disulfide intercalating cross-linking reagent.

Specifications

Molecular Weight (g/mol) 222.76 g/mol
CAS 106139-15-5
Content And Storage 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture)
Shipping Condition Room temperature
Quantity 200 mg
Synonym (S)-2-Pyridylthio Cysteamine Hydrochloride
Solubility H2O : 50 mg/mL (224.46 mM; Need ultrasonic) ∣ DMSO : ≥ 32 mg/mL (143.65 mM)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.