missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorocinnamic acid, predominantly trans, 98%, Thermo Scientific™

Product Code. 11381917
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381917 1 g 1g
11391917 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381917 Supplier Thermo Scientific Alfa Aesar Supplier No. L07468.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392-22-3
Molecular Formula C9H6ClFO2
Molecular Weight (g/mol) 200.593
MDL Number MFCD00051582
InChI Key NDWALECYVLNBQG-SNAWJCMRSA-N
Synonym 2-chloro-6-fluorocinnamic acid, 3-2-chloro-6-fluorophenyl acrylic acid, 2e-3-2-chloro-6-fluorophenyl prop-2-enoic acid, 2e-3-2-chloro-6-fluorophenyl acrylic acid, e-3-2-chloro-6-fluoro-phenyl-acrylic acid, 2-propenoic acid, 3-2-chloro-6-fluorophenyl-, 2e, 3-2-chloro-6-fluorophenyl prop-2-enoic acid, pubchem3430, rarechem bk hw 0069
PubChem CID 5324668
IUPAC Name (E)-3-(2-chloro-6-fluorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C(=C1)Cl)C=CC(=O)O)F

Specifications

Melting Point 164°C to 172°C
Quantity 1 g
Beilstein 3129946
Formula Weight 200.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-fluorocinnamic acid, predominantly trans
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.