CAS RN 392-22-3
IUPAC Name:
(2E)-3-(2-chloro-6-fluorophenyl)prop-2-enoate
Synonyms:
(2E)-3-(2-chloro-6-fluorophenyl)prop-2-enoate
Molecular Weight (g/mol):
199.59
Molecular Formula:
C9H5ClFO2
InChi Key:
NDWALECYVLNBQG-SNAWJCMRSA-M
SMILES:
[O-]C(=O)\C=C\C1=C(F)C=CC=C1Cl
1
–
1
of
1
results
1
2-Chloro-6-fluorocinnamic acid, predominantly trans, 98%, Thermo Scientific™
CAS: 392-22-3 Molecular Formula: C9H6ClFO2 Molecular Weight (g/mol): 200.593 MDL Number: MFCD00051582 InChI Key: NDWALECYVLNBQG-SNAWJCMRSA-N Synonym: 2-chloro-6-fluorocinnamic acid,3-2-chloro-6-fluorophenyl acrylic acid,2e-3-2-chloro-6-fluorophenyl prop-2-enoic acid,2e-3-2-chloro-6-fluorophenyl acrylic acid,e-3-2-chloro-6-fluoro-phenyl-acrylic acid,2-propenoic acid, 3-2-chloro-6-fluorophenyl-, 2e,3-2-chloro-6-fluorophenyl prop-2-enoic acid,pubchem3430,rarechem bk hw 0069 PubChem CID: 5324668 IUPAC Name: (E)-3-(2-chloro-6-fluorophenyl)prop-2-enoic acid SMILES: C1=CC(=C(C(=C1)Cl)C=CC(=O)O)F