missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ 18/10 Stainless Steel Spatulas, With knob
Manufactured from 18/10 stainless steel
Brand: Bochem™ 3272
Additional Details : Weight : 0.10000kg
Description
- Made from 18/10 stainless steel
Specifications
1 Ea. | |
Spatula | |
11 mm | |
Stainless Steel (18/10) | |
300 x 16 mm | |
300 mm |
With Knob | |
70 mm | |
11 mm | |
5mm | |
Stainless Steel (18/10) |