Learn More
Hamilton™ 1700 Series Standard PTFE Instrument Syringe
Setting the standard for quality and reliability.
£122.00 - £171.00
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10490405
|
Hamilton™
5495-22 |
100 μL |
£168.00
Each |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10097109
|
Hamilton™
5495-10 |
12.5 μL |
£171.00
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11580112
|
Hamilton™
5495-30 |
250 μL |
£122.00
Each |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Syringe life is exceptional when used in conjunction with Hamilton™ precision syringe drives
- Enables precision liquid handling
- Gastight for leak-free performance