Learn More
Hamilton™ 1700 Series Standard PTFE Instrument Syringe
Product Code. 11580112
Click to view available options
Volume (Metric):
12.5 μL
100 μL
250 μL
Unit Size:
Each
Product Code. 11580112
Brand: Hamilton™ 549530
Setting the standard for quality and reliability.
  • Syringe life is exceptional when used in conjunction with Hamilton™ precision syringe drives
  • Enables precision liquid handling
  • Gastight for leak-free performance
Specifications
Volume (Metric) 250 μL